3-Methyl-3,4-dihydro-2H-1,4-benzoxazine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine

044112-500MG 044112-1G 044112-5G
101864-208EA 344.62 USD
101864-208 101864-210 101864-206
3-Methyl-3,4-dihydro-2H-1,4-benzoxazine ≥95%
3-Methyl-3,4-dihydro-2H-1,4-benzoxazine

Hazard: Irritant

Formula: C₉H₁₁NO
MW: 149.19 g/mol
Boiling Pt: 98…100 °C
MDL Number: MFCD01935996
CAS Number: 32329-20-7

Order Now

Specification Test Results

Melting point 98-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR