1-(3,5-Dichlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
044097-1G
044097-500MG
044097-5G
101864-120EA
1109.23
USD
101864-120
101864-122
101864-124
1-(3,5-Dichlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylic acid ≥95%
1-(3,5-Dichlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₇Cl₂N₃O₂ Melting Pt: 202…204 °C |
MDL Number:
MFCD00173546 CAS Number: 338408-13-2 |
Specification Test Results
Melting point | 202-204°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...