5-Chloro-4-(4-chlorophenyl)-1,2,3-thiadiazole ≥95%

Supplier: Matrix Scientific

044096-5G 044096-1G 044096-500MG
101864-114EA 375.38 USD
101864-114 101864-116 101864-118
5-Chloro-4-(4-chlorophenyl)-1,2,3-thiadiazole ≥95%
5-Chloro-4-(4-chlorophenyl)-1,2,3-thiadiazole

Hazard: Irritant

Formula: C₈H₄Cl₂N₂S
Melting Pt: 103…104 °C
MDL Number: MFCD00173533
CAS Number: 338407-87-7

Order Now

Specification Test Results

Melting point 103-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR