5-Chloro-4-(2,4-dichlorophenyl)-1,2,3-thiadiazole ≥95%

Supplier: Matrix Scientific

044095-1G 044095-5G 044095-500MG
101864-108EA 280.6 USD
101864-108 101864-110 101864-112
5-Chloro-4-(2,4-dichlorophenyl)-1,2,3-thiadiazole ≥95%
5-Chloro-4-(2,4-dichlorophenyl)-1,2,3-thiadiazole

Hazard: Irritant

Formula: C₈H₃Cl₃N₂S
Melting Pt: 72…74 °C
MDL Number: MFCD00173532
CAS Number: 338407-85-5

Order Now

Specification Test Results

Melting point 72-74°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR