3-(Hydroxymethyl)-5-methyl-N-(1,3-thiazol-2-yl)-4-isoxazolecarboxamide ≥95%
Supplier: Matrix Scientific
044090-1G
044090-500MG
101864-082EA
1855.38
USD
101864-082
101864-080
3-(Hydroxymethyl)-5-methyl-N-(1,3-thiazol-2-yl)-4-isoxazolecarboxamide ≥95%
3-(Hydroxymethyl)-5-methyl-N-(1,3-thiazol-2-yl)-4-isoxazolecarboxamide
Hazard: Irritant
Formula:
C₉H₉N₃O₃S Melting Pt: 230…232 °C |
MDL Number:
MFCD00664755 CAS Number: 338408-93-8 |
Specification Test Results
Melting point | 230-232°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...