Ethyl-4-methyl-2-(2-thienyl)-1,3-thiazole-5-carboxylate ≥95%
Supplier: Matrix Scientific
044084-500MG
044084-5G
044084-1G
101864-050EA
698.46
USD
101864-050
101864-052
101864-048
Ethyl-4-methyl-2-(2-thienyl)-1,3-thiazole-5-carboxylate ≥95%
Ethyl-4-methyl-2-(2-thienyl)-1,3-thiazole-5-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₁NO₂S₂ Melting Pt: 66…68 °C |
MDL Number:
MFCD00208019 CAS Number: 56421-62-6 |
Specification Test Results
Melting point | 66-68°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...