(2-Methyl-3-phenyltetrahydro-5-isoxazolyl)methanol ≥95%

Supplier: Matrix Scientific

044070-1G 044070-500MG
101863-978EA 927.69 USD
101863-978 101863-976
(2-Methyl-3-phenyltetrahydro-5-isoxazolyl)methanol ≥95%
(2-Methyl-3-phenyltetrahydro-5-isoxazolyl)methanol

Hazard: Irritant

Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
Melting Pt: 78…84 °C
MDL Number: MFCD00171848
CAS Number: 19345-02-9

Order Now

Specification Test Results

Melting point 78-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR