5-Methyl-3-phenyl-4-isoxazolecarbohydrazide ≥95%

Supplier: Matrix Scientific

044063-1G 044063-500MG 044063-5G
101863-948EA 1073.85 USD
101863-948 101863-946 101863-944
5-Methyl-3-phenyl-4-isoxazolecarbohydrazide ≥95%
5-Methyl-3-phenyl-4-isoxazolecarbohydrazide

Hazard: Irritant

Formula: C₁₁H₁₁N₃O₂
Melting Pt: 173…175 °C
MDL Number: MFCD00119108
CAS Number: 18336-75-9

Order Now

Specification Test Results

Melting point 173-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR