5-(Carboxymethyl)-1-(4-methoxyphenyl)-1H-1,2,3-triazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

044047-500MG 044047-1G
101863-852EA 556.92 USD
101863-852 101863-854
5-(Carboxymethyl)-1-(4-methoxyphenyl)-1H-1,2,3-triazole-4-carboxylic acid ≥95%
5-(Carboxymethyl)-1-(4-methoxyphenyl)-1H-1,2,3-triazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₁N₃O₅
Melting Pt: 200…201 °C
MDL Number: MFCD05021856
CAS Number: 439095-14-4

Order Now

Specification Test Results

Melting point 200-201°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR