3-[4-(3-Chlorophenoxy)-3-nitrophenyl]acrylic acid ≥95%
Supplier: Matrix Scientific
044042-1G
044042-500MG
044042-5G
101863-828EA
747.69
USD
101863-828
101863-830
101863-832
3-[4-(3-Chlorophenoxy)-3-nitrophenyl]acrylic acid ≥95%
3-[4-(3-Chlorophenoxy)-3-nitrophenyl]acrylic acid
Hazard: Irritant
Formula:
C₁₅H₁₀ClNO₅ Melting Pt: 164…165 °C |
MDL Number:
MFCD00138698 CAS Number: 338403-64-8 |
Specification Test Results
Melting point | 164-165°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...