4-[(3,5-Dimethyl-1H-pyrazol-4-yl)methyl]benzenol ≥95%
Supplier: Matrix Scientific
044034-500MG
044034-1G
044034-5G
101863-782EA
232.31
USD
101863-782
101863-780
101863-784
4-[(3,5-Dimethyl-1H-pyrazol-4-yl)methyl]benzenol ≥95%
4-[(3,5-Dimethyl-1H-pyrazol-4-yl)methyl]benzenol
Hazard: Irritant
Formula:
C₁₂H₁₄N₂O Melting Pt: 232…234 °C |
MDL Number:
MFCD00138769 CAS Number: 75998-99-1 |
Specification Test Results
Melting point | 232-234°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...