Ethyl-2-(4-bromophenyl)-1-indolizinecarboxylate ≥95%

Supplier: Matrix Scientific

044028-5G 044028-1G 044028-500MG
101863-752EA 556.92 USD
101863-752 101863-756 101863-754
Ethyl-2-(4-bromophenyl)-1-indolizinecarboxylate ≥95%
Ethyl-2-(4-bromophenyl)-1-indolizinecarboxylate

Hazard: Irritant

Formula: C₁₇H₁₄BrNO₂
Melting Pt: 124…126 °C
MDL Number: MFCD07366544
CAS Number: 885949-62-2

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR