2-Oxo-1,2-dihydro-3-pyridinecarbaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Oxo-1,2-dihydropyridine-3-carbaldehyde
044020-5G
044020-1G
101863-714EA
161.54
USD
101863-714
101863-712
2-Oxo-1,2-dihydro-3-pyridinecarbaldehyde ≥95%
2-Oxo-1,2-dihydro-3-pyridinecarbaldehyde
Hazard: Irritant
Formula:
C₆H₅NO₂ MW: 123.11 g/mol Melting Pt: 220…222 °C |
MDL Number:
MFCD07698588 CAS Number: 36404-89-4 |
Specification Test Results
Melting point | 220-222°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...