5-[(4-Bromophenyl)sulfanyl]-2-thiophenecarbaldehyde ≥95%

Supplier: Matrix Scientific

044019-500MG 044019-1G 044019-5G
101863-706EA 181.54 USD
101863-706 101863-708 101863-710
5-[(4-Bromophenyl)sulfanyl]-2-thiophenecarbaldehyde ≥95%
5-[(4-Bromophenyl)sulfanyl]-2-thiophenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₇BrOS₂
Melting Pt: 57…59 °C
MDL Number: MFCD07698587
CAS Number: 886361-55-3

Order Now

Specification Test Results

Melting point 57-59°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR