5-[(4-Chlorophenyl)sulfanyl]-2-thiophenecarbaldehyde ≥95%

Supplier: Matrix Scientific

044018-1G 044018-5G 044018-500MG
101863-704EA 602.6 USD
101863-704 101863-702 101863-700
5-[(4-Chlorophenyl)sulfanyl]-2-thiophenecarbaldehyde ≥95%
5-[(4-Chlorophenyl)sulfanyl]-2-thiophenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₇ClOS₂
Melting Pt: 40…42 °C
MDL Number: MFCD07698586
CAS Number: 139120-69-7

Order Now

Specification Test Results

Melting point 40-42°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR