5-(1-Pyrrolidinyl)-2-thiophenecarbaldehyde ≥95%
Supplier: Matrix Scientific
044012-1G
044012-500MG
101863-672EA
375.38
USD
101863-672
101863-674
5-(1-Pyrrolidinyl)-2-thiophenecarbaldehyde ≥95%
5-(1-Pyrrolidinyl)-2-thiophenecarbaldehyde
Hazard: Irritant
Formula:
C₉H₁₁NOS Melting Pt: 103…106 °C |
MDL Number:
MFCD07440065 CAS Number: 24372-47-2 |
Specification Test Results
Melting point | 103-106°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...