2-(4-Hydroxyphenyl)-3-methyl-4-quinolinecarboxylic acid ≥95%
Supplier: Matrix Scientific
043998-1G
043998-5G
043998-500MG
101863-604EA
532.31
USD
101863-604
101863-602
101863-600
2-(4-Hydroxyphenyl)-3-methyl-4-quinolinecarboxylic acid ≥95%
2-(4-Hydroxyphenyl)-3-methyl-4-quinolinecarboxylic acid
Hazard: Irritant
Formula:
C₁₇H₁₃NO₃ MW: 279.3 g/mol Melting Pt: 338 °C |
MDL Number:
MFCD06200949 CAS Number: 107419-49-8 |
Specification Test Results
Melting point | 338°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...