3-(2-Amino[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)propyl acetate ≥95%
Supplier: Matrix Scientific
043983-500MG
043983-5G
043983-1G
101863-518EA
556.92
USD
101863-518
101863-522
101863-520
3-(2-Amino[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)propyl acetate ≥95%
3-(2-Amino[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)propyl acetate
Hazard: Irritant
Formula:
C₁₀H₁₃N₅O₂ Melting Pt: 150 °C |
MDL Number:
MFCD06659547 CAS Number: 85599-38-8 |
Specification Test Results
Melting point | 150°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...