Methyl-4'-amino[1,1'-biphenyl]-4-carboxylate ≥95%
Supplier: Matrix Scientific
043977-1G
043977-500MG
043977-5G
101863-492EA
1600
USD
101863-492
101863-490
101863-488
Methyl-4'-amino[1,1'-biphenyl]-4-carboxylate ≥95%
Methyl-4'-amino[1,1'-biphenyl]-4-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₃NO₂ Melting Pt: 179…182 °C |
MDL Number:
MFCD02628073 CAS Number: 5730-76-7 |
Specification Test Results
Melting point | 179-182°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...