N-Hydroxy-1-hydrazinecarboxamide ≥95%
Supplier: Matrix Scientific
Synonyms:
N-Hydroxyhydrazinecarboxamide
043952-500MG
043952-5G
043952-1G
101863-374EA
307.69
USD
101863-374
101863-378
101863-376
N-Hydroxy-1-hydrazinecarboxamide ≥95%
N-Hydroxy-1-hydrazinecarboxamide
Hazard: Irritant
Formula:
CH₅N₃O₂ MW: 91.07 g/mol Melting Pt: 130…132 °C |
MDL Number:
MFCD03791199 CAS Number: 21520-79-6 |
Specification Test Results
Melting point | 130-132°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...