Ethyl-6-methyl-2-oxo-4-phenyl-1,2-dihydro-5-pyrimidinecarboxylate ≥95%
Supplier: Matrix Scientific
043947-1G
043947-5G
043947-500MG
101863-346EA
350.77
USD
101863-346
101863-344
101863-348
Ethyl-6-methyl-2-oxo-4-phenyl-1,2-dihydro-5-pyrimidinecarboxylate ≥95%
Ethyl-6-methyl-2-oxo-4-phenyl-1,2-dihydro-5-pyrimidinecarboxylate
Hazard: Irritant
Formula:
C₁₄H₁₄N₂O₃ Melting Pt: 192…194 °C |
MDL Number:
MFCD03791196 CAS Number: 69207-36-9 |
Specification Test Results
Melting point | 192-194°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...