4-(2-Hydroxyethyl)-2,5-dimethyl-1,2-dihydro-3H-pyrazol-3-one ≥95%
Supplier: Matrix Scientific
Synonyms:
1,2-Dihydro-2,5-dimethyl-4-(2-hydroxyethyl)-3H-pyrazol-3-one
043938-5G
043938-500MG
043938-1G
101863-294EA
438.46
USD
101863-294
101863-292
101863-290
4-(2-Hydroxyethyl)-2,5-dimethyl-1,2-dihydro-3H-pyrazol-3-one ≥95%
4-(2-Hydroxyethyl)-2,5-dimethyl-1,2-dihydro-3H-pyrazol-3-one
Hazard: Irritant
Formula:
C₇H₁₂N₂O₂ MW: 156.19 g/mol Melting Pt: 78…81 °C |
MDL Number:
MFCD03617955 CAS Number: 258281-02-6 |
Specification Test Results
Melting point | 78-81°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...