1-(6,7-Dichloro-8-nitro-2,3-dihydro-1,4-benzodioxin-5-yl)-1-ethanone ≥95%
Supplier: Matrix Scientific
043937-5G
043937-500MG
043937-1G
101863-286EA
467.69
USD
101863-286
101863-284
101863-288
1-(6,7-Dichloro-8-nitro-2,3-dihydro-1,4-benzodioxin-5-yl)-1-ethanone ≥95%
1-(6,7-Dichloro-8-nitro-2,3-dihydro-1,4-benzodioxin-5-yl)-1-ethanone
Hazard: Irritant
Formula:
C₁₀H₇Cl₂NO₅ Melting Pt: 182…185 °C |
MDL Number:
MFCD03791191 CAS Number: 166816-12-2 |
Specification Test Results
Melting point | 182-185°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...