Dimethyl-2-(2,4-dinitrophenyl)-2-methylmalonate ≥95%
Supplier: Matrix Scientific
043936-5G
043936-1G
043936-500MG
101863-282EA
602.6
USD
101863-282
101863-280
101863-278
Dimethyl-2-(2,4-dinitrophenyl)-2-methylmalonate ≥95%
Dimethyl-2-(2,4-dinitrophenyl)-2-methylmalonate
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O₈ Melting Pt: 101…103 °C |
MDL Number:
MFCD03617951 CAS Number: 861209-84-9 |
Specification Test Results
Melting point | 101-103°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...