4-(3-Chloro-2-thienyl)-6-methyl-2-pyrimidinamine ≥95%

Supplier: Matrix Scientific

043931-500MG
101863-264EA 556.92 USD
101863-264
4-(3-Chloro-2-thienyl)-6-methyl-2-pyrimidinamine ≥95%
4-(3-Chloro-2-thienyl)-6-methyl-2-pyrimidinamine

Hazard: Irritant

Formula: C₉H₈ClN₃S
Melting Pt: 163…164 °C
MDL Number: MFCD03791190
CAS Number: 863305-81-1

Order Now

Specification Test Results

Melting point 163-164°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR