Methyl-3-(4-formyl-2-nitrophenoxy)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

043918-1G 043918-5G 043918-500MG
101863-198EA 350.77 USD
101863-198 101863-196 101863-200
Methyl-3-(4-formyl-2-nitrophenoxy)benzenecarboxylate ≥95%
Methyl-3-(4-formyl-2-nitrophenoxy)benzenecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₁NO₆
Melting Pt: 112…113 °C
MDL Number: MFCD03425737
CAS Number: 439095-08-6

Order Now

Specification Test Results

Melting point 112-113°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR