2-[3-(Trifluoromethyl)benzyl]-1,3-thiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

043914-500MG 043914-5G 043914-1G
101863-182EA 1591.6 USD
101863-182 101863-178 101863-180
2-[3-(Trifluoromethyl)benzyl]-1,3-thiazole-4-carboxylic acid ≥95%
2-[3-(Trifluoromethyl)benzyl]-1,3-thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₈F₃NO₂S
Melting Pt: 123…125 °C
MDL Number: MFCD02186742
CAS Number: 478030-66-9

Order Now

Specification Test Results

Melting point 123-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR