2-Thioxo-1,2-dihydro-3H-indol-3-one-N-(2,4,6-trichlorophenyl)hydrazone ≥95%
Supplier: Matrix Scientific
043909-5G
043909-1G
043909-500MG
101863-162EA
375.38
USD
101863-162
101863-166
101863-164
2-Thioxo-1,2-dihydro-3H-indol-3-one-N-(2,4,6-trichlorophenyl)hydrazone ≥95%
2-Thioxo-1,2-dihydro-3H-indol-3-one-N-(2,4,6-trichlorophenyl)hydrazone
Hazard: Irritant
Formula:
C₁₄H₈Cl₃N₃S Melting Pt: 254…255 °C |
MDL Number:
MFCD06200943 CAS Number: 862665-28-9 |
Specification Test Results
Melting point | 254-255°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...