2-Benzyl-3-hydrazinoquinoxaline ≥95%

Supplier: Matrix Scientific

043896-500MG
101863-104EA 688.85 USD
101863-104
2-Benzyl-3-hydrazinoquinoxaline ≥95%
2-Benzyl-3-hydrazinoquinoxaline

Hazard: Irritant

Formula: C₁₅H₁₄N₄
Melting Pt: 153…155 °C
MDL Number: MFCD08056617
CAS Number: 223929-23-5

Order Now

Specification Test Results

Melting point 153-155°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR