Ethyl-2-(4-methylphenyl)-1-indolizinecarboxylate ≥95%

Supplier: Matrix Scientific

043894-500MG
101863-096EA 1076.92 USD
101863-096
Ethyl-2-(4-methylphenyl)-1-indolizinecarboxylate ≥95%
Ethyl-2-(4-methylphenyl)-1-indolizinecarboxylate

Hazard: Irritant

Formula: C₁₈H₁₇NO₂
Melting Pt: 119…121 °C
MDL Number: MFCD02185989
CAS Number: 449778-84-1

Order Now

Specification Test Results

Melting point 119-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR