5-Aminobenzimidazole ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Amino-1H-benzimidazole
043881-5G
043881-1G
101863-040EA
32.31
USD
101863-040
101863-042
5-Aminobenzimidazole ≥95%
5-Aminobenzimidazole
Hazard: Irritant
Formula:
C₇H₇N₃ MW: 133.15 g/mol Melting Pt: 163…165 °C |
MDL Number:
MFCD00465258 CAS Number: 934-22-5 |
Specification Test Results
Melting point | 163-165°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...