6-Nitrobenzoxazole ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Nitrobenzo[d]oxazole

043877-5G 043877-1G
101863-022EA 43.08 USD
101863-022 101863-024
6-Nitrobenzoxazole ≥95%
6-Nitrobenzoxazole

Hazard: Irritant

Formula: C₇H₄N₂O₃
MW: 164.1183 g/mol
Melting Pt: 150…152 °C
MDL Number: MFCD00797933
CAS Number: 17200-30-5

Order Now

Specification Test Results

Melting point 150-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR