2-(3,5-Dimethylphenoxy)benzenecarbaldehyde oxime ≥95%
Supplier: Matrix Scientific
043874-1G
043874-500MG
101863-008EA
1400
USD
101863-008
101863-006
2-(3,5-Dimethylphenoxy)benzenecarbaldehyde oxime ≥95%
2-(3,5-Dimethylphenoxy)benzenecarbaldehyde oxime
Hazard: Irritant
Formula:
C₁₅H₁₅NO₂ Melting Pt: 109…111 °C |
MDL Number:
MFCD02091013 CAS Number: 449778-77-2 |
Specification Test Results
Melting point | 109-111°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...