Ethyl-2-amino-4-(2-naphthyl)-1H-pyrrole-3-carboxylate ≥95%
Supplier: Matrix Scientific
043862-1G
043862-500MG
101862-960EA
1483.08
USD
101862-960
101862-962
Ethyl-2-amino-4-(2-naphthyl)-1H-pyrrole-3-carboxylate ≥95%
Ethyl-2-amino-4-(2-naphthyl)-1H-pyrrole-3-carboxylate
Hazard: Irritant
Formula:
C₁₇H₁₆N₂O₂ Melting Pt: 154…156 °C |
MDL Number:
MFCD00974201 CAS Number: 338400-98-9 |
Specification Test Results
Melting point | 154-156°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...