Methyl-2-[(4-formyl-1-methyl-3-phenyl-1H-pyrazol-5-yl)sulfanyl]benzenecarboxylate ≥95%
Supplier: Matrix Scientific
043845-1G
043845-500MG
043845-5G
101862-870EA
921.54
USD
101862-870
101862-872
101862-874
Methyl-2-[(4-formyl-1-methyl-3-phenyl-1H-pyrazol-5-yl)sulfanyl]benzenecarboxylate ≥95%
Methyl-2-[(4-formyl-1-methyl-3-phenyl-1H-pyrazol-5-yl)sulfanyl]benzenecarboxylate
Hazard: Irritant
Formula:
C₁₉H₁₆N₂O₃S Melting Pt: 95…98 °C |
MDL Number:
MFCD00244902 CAS Number: 321538-23-2 |
Specification Test Results
Melting point | 95-98°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...