5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde ≥95%
Supplier: Matrix Scientific
043842-1G
043842-500MG
043842-5G
101862-854EA
652.31
USD
101862-854
101862-856
101862-852
5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde ≥95%
5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde
Hazard: Irritant
Formula:
C₁₁H₉ClN₂O Melting Pt: 62…64 °C |
MDL Number:
MFCD00244899 CAS Number: 883-38-5 |
Specification Test Results
Melting point | 62-64°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...