2-[3-(Dimethylamino)-1-methoxy-2-propenylidene]malononitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(3-(Dimethylamino)-1-methoxyallylidene)malononitrile
043837-1G
043837-5G
043837-500MG
101862-826EA
278.46
USD
101862-826
101862-828
101862-830
2-[3-(Dimethylamino)-1-methoxy-2-propenylidene]malononitrile ≥95%
2-[3-(Dimethylamino)-1-methoxy-2-propenylidene]malononitrile
Hazard: Irritant
Formula:
C₉H₁₁N₃O MW: 177.21 g/mol Melting Pt: 130 °C |
MDL Number:
MFCD00975440 CAS Number: 95689-38-6 |
Specification Test Results
Melting point | 130°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...