3-(4-Chlorophenyl)-2,5-furandione ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(4-Chlorophenyl)-2,5-dihydrofuran-2,5-dione

043833-5G 043833-1G 043833-500MG
101862-804EA 232.31 USD
101862-804 101862-806 101862-802
3-(4-Chlorophenyl)-2,5-furandione ≥95%
3-(4-Chlorophenyl)-2,5-furandione

Hazard: Irritant

Formula: C₁₀H₅ClO₃
MW: 208.6 g/mol
Melting Pt: 152…154 °C
MDL Number: MFCD00974639
CAS Number: 3152-15-6

Order Now

Specification Test Results

Melting point 152-154°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR