Ethyl-2-[(4-chlorobenzoyl)amino]acetate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-[(4-chlorophenyl)formamido]acetate
043832-500MG
043832-1G
043832-5G
101862-800EA
532.31
USD
101862-800
101862-796
101862-798
Ethyl-2-[(4-chlorobenzoyl)amino]acetate ≥95%
Ethyl-2-[(4-chlorobenzoyl)amino]acetate
Hazard: Irritant
Formula:
C₁₁H₁₂ClNO₃ MW: 241.68 g/mol Melting Pt: 118…120 °C |
MDL Number:
MFCD00095955 CAS Number: 39735-52-9 |
Specification Test Results
Melting point | 118-120°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...