1-(3-Methoxypropyl)-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile ≥95%
Supplier: Matrix Scientific
043829-1G
043829-500MG
043829-5G
101862-780EA
927.69
USD
101862-780
101862-782
101862-778
1-(3-Methoxypropyl)-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile ≥95%
1-(3-Methoxypropyl)-2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinecarbonitrile
Hazard: Irritant
Formula:
C₉H₁₁N₃O₃ Melting Pt: 156…157 °C |
MDL Number:
MFCD00172961 CAS Number: 338399-18-1 |
Specification Test Results
Melting point | 156-157°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...