5-Methyl-2-(3-pyridinyl)-1,3-thiazol-4-ol ≥95%

Supplier: Matrix Scientific

043826-1G 043826-500MG 043826-5G
101862-760EA 149.5 USD
101862-760 101862-762 101862-764
5-Methyl-2-(3-pyridinyl)-1,3-thiazol-4-ol ≥95%
5-Methyl-2-(3-pyridinyl)-1,3-thiazol-4-ol

Hazard: Irritant

Formula: C₉H₈N₂OS
Melting Pt: 202…205 °C
MDL Number: MFCD00172527
CAS Number: 131786-48-6

Order Now

Specification Test Results

Melting point 202-205°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR