5-Chloro-4-phenyl-1,2,3-thiadiazole ≥95%

Supplier: Matrix Scientific

043823-500MG 043823-5G 043823-1G
101862-744EA 467.69 USD
101862-744 101862-746 101862-742
5-Chloro-4-phenyl-1,2,3-thiadiazole ≥95%
5-Chloro-4-phenyl-1,2,3-thiadiazole

Hazard: Irritant

Formula: C₈H₅ClN₂S
Melting Pt: 93…95 °C
MDL Number: MFCD00100225
CAS Number: 53646-00-7

Order Now

Specification Test Results

Melting point 93-95°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR