Ethyl-2-(acetylamino)-2-[2-(2,5-dichlorophenyl)hydrazono]acetate ≥95%
Supplier: Matrix Scientific
043822-500MG
043822-5G
043822-1G
101862-738EA
467.69
USD
101862-738
101862-736
101862-740
Ethyl-2-(acetylamino)-2-[2-(2,5-dichlorophenyl)hydrazono]acetate ≥95%
Ethyl-2-(acetylamino)-2-[2-(2,5-dichlorophenyl)hydrazono]acetate
Hazard: Irritant
Formula:
C₁₂H₁₃Cl₂N₃O₃ Melting Pt: 190…192 °C |
MDL Number:
MFCD08056615 CAS Number: 886361-49-5 |
Specification Test Results
Melting point | 190-192°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...