Ethyl-4-methyl-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-methyl-2-(2-pyridyl)thiazole-5-carboxylate

043803-1G 043803-500MG 043803-5G
101862-646EA 802.7 USD
101862-646 101862-642 101862-644
Ethyl-4-methyl-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate ≥95%
Ethyl 4-methyl-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂S
Melting Pt: 82…84 °C
MDL Number: MFCD01936157
CAS Number: 383146-88-1

Order Now

Specification Test Results

Melting point 82-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR