Ethyl-5-phenyl-2-(1-pyrrolidinyl)-1H-pyrrole-3-carboxylate ≥95%
Supplier: Matrix Scientific
043798-1G
043798-5G
043798-500MG
101862-616EA
250.77
USD
101862-616
101862-618
101862-614
Ethyl-5-phenyl-2-(1-pyrrolidinyl)-1H-pyrrole-3-carboxylate ≥95%
Ethyl-5-phenyl-2-(1-pyrrolidinyl)-1H-pyrrole-3-carboxylate
Hazard: Irritant
Formula:
C₁₇H₂₀N₂O₂ Melting Pt: 146 °C |
MDL Number:
MFCD00171771 CAS Number: 118082-59-0 |
Specification Test Results
Melting point | 167-169°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...