Methyl-4-hydroxy-1-(4-methoxyphenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylate ≥95%

Supplier: Matrix Scientific

043783-5G 043783-1G 043783-500MG
101862-540EA 1073.85 USD
101862-540 101862-536 101862-538
Methyl-4-hydroxy-1-(4-methoxyphenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylate ≥95%
Methyl-4-hydroxy-1-(4-methoxyphenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₅
Melting Pt: 184…186 °C
MDL Number: MFCD00141150
CAS Number: 121582-61-4

Order Now

Specification Test Results

Melting point 184-186°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR