Dimethyl-2-[2-(4-methoxyphenyl)hydrazono]-3-oxopentanedioate ≥95%
Supplier: Matrix Scientific
043781-5G
043781-1G
043781-500MG
101862-530EA
532.31
USD
101862-530
101862-528
101862-526
Dimethyl-2-[2-(4-methoxyphenyl)hydrazono]-3-oxopentanedioate ≥95%
Dimethyl-2-[2-(4-methoxyphenyl)hydrazono]-3-oxopentanedioate
Hazard: Irritant
Formula:
C₁₄H₁₆N₂O₆ Melting Pt: 110…113 °C |
MDL Number:
MFCD07698581 CAS Number: 121582-52-3 |
Specification Test Results
Melting point | 110-113°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...