Ethyl-3-(benzoylamino)-2-oxo-6-phenyl-2H-pyran-5-carboxylate ≥95%

Supplier: Matrix Scientific

043779-500MG 043779-1G 043779-5G
101862-518EA 602.6 USD
101862-518 101862-514 101862-516
Ethyl-3-(benzoylamino)-2-oxo-6-phenyl-2H-pyran-5-carboxylate ≥95%
Ethyl-3-(benzoylamino)-2-oxo-6-phenyl-2H-pyran-5-carboxylate

Hazard: Irritant

Formula: C₂₁H₁₇NO₅
Melting Pt: 147…149 °C
MDL Number: MFCD00139773
CAS Number: 127143-18-4

Order Now

Specification Test Results

Melting point 147-149°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR