(2,4-Dichlorophenoxy)thioacetamide ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(2,4-Dichlorophenoxy)thioacetamide
043774-1G
043774-500MG
043774-5G
101862-496EA
278.46
USD
101862-496
101862-498
101862-500
(2,4-Dichlorophenoxy)thioacetamide ≥95%
(2,4-Dichlorophenoxy)thioacetamide
Hazard: Irritant
Formula:
C₈H₇Cl₂NOS MW: 236.11 Melting Pt: 139…141 °C |
MDL Number:
MFCD00172128 CAS Number: 2302-32-1 |
Specification Test Results
Melting point | 139-141°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...