2-(5-Bromo-1H-indol-3-yl)-1-ethanol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(5-Bromo-1H-indol-3-yl)ethanol
, 5-Bromoindole-3-ethanol
, 5-Bromo-3-(2-hydroxyethyl)indole
043763-5G
043763-1G
101862-452EA
92.31
USD
101862-452
101862-454
2-(5-Bromo-1H-indol-3-yl)-1-ethanol ≥95%
2-(5-Bromo-1H-indol-3-yl)-1-ethanol
Hazard: Irritant
Formula:
C₁₀H₁₀BrNO MW: 240.11 g/mol Melting Pt: 92…94 °C |
MDL Number:
MFCD00130167 CAS Number: 32774-29-1 |
Specification Test Results
Melting point | 92-94°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...